Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081241.2 | NP_001074710.2 | 12, 13 | 1219 | ||
GenBank mRNA | AK052316.1 | - | 12, 13 | 1168 | ||
AK122573.1 | - | 12, 13 | 1193 | |||
BC030451.1 | - | 2, 3 | 282 | 5400266 | ||
BC079880.1 | - | 12, 13 | 1133 | 6852251 |