Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029420.4 | NP_083696.2 | 6 | 1161 | ||
GenBank mRNA | AK010748.1 | - | 6 | 1161 | ||
AK010828.1 | - | 5 | 1230 | |||
AK014753.1 | - | 5 | 1283 | |||
AK049139.1 | - | 6 | 1162 | |||
AK053901.1 | - | 5 | 1051 | |||
BC055909.1 | - | 6 | 1112 | |||
BC145468.1 | - | 5 | 973 | |||
BC145688.1 | - | 6 | 1026 | 40130991 |