Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cant1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025617.2 | NP_001020788.1 | 3 | 870 | ||
NM_001025618.2 | NP_001020789.1 | 3 | 828 | |||
NM_001267591.1 | NP_001254520.1 | 2 | 671 | |||
NM_001267592.1 | NP_001254521.1 | 3 | 870 | |||
NM_029502.3 | NP_083778.2 | 3 | 1022 | |||
GenBank mRNA | AK006565.1 | - | 3 | 721 | ||
AK017942.1 | - | 3 | 740 | |||
AK049879.1 | - | 3 | 764 | |||
AK081118.1 | - | 3 | 927 | |||
AK085059.1 | - | 2 | 2118 | |||
AK166384.1 | - | 2 | 2032 | |||
BC020003.1 | - | 2 | 670 | 4208801 | ||
CA320376.1 | - | 4 | 558 |