Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACRV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001612.5 | NP_001603.1 | 4 | 820 | ||
NM_020069.4 | NP_064454.1 | 4 | 763 | |||
NM_020107.4 | NP_064492.1 | 5 | 655 | |||
NM_020108.4 | NP_064493.1 | 5 | 610 | |||
GenBank mRNA | AB451476.1 | - | 4 | 642 | ||
AK223335.1 | - | 4 | 769 | |||
BC014588.2 | - | 4 | 796 | 4825974 | ||
BG720361.1 | - | 5 | 580 | |||
M82967.1 | - | 4 | 697 | |||
M82968.1 | - | 4 | 754 |