Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029570.3 | NP_083846.2 | 5, 6 | 680 | ||
GenBank mRNA | AK129248.1 | - | 5, 6 | 672 | ||
AK154156.1 | - | 5, 6 | 684 | |||
AK155372.1 | - | 5, 6 | 684 | |||
BC076603.1 | - | 5, 6 | 550 | 30629594 | ||
CN535194.1 | - | 5, 6 | 680 |