Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gid8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289651.1 | NP_001276580.1 | 5 | 806 | ||
NM_001289652.1 | NP_001276581.1 | 5 | 772 | |||
NM_029607.2 | NP_083883.1 | 4 | 1292 | |||
GenBank mRNA | AK009106.1 | - | 5 | 732 | ||
AK213539.1 | - | 1 | 71 | |||
BC052539.1 | - | 5 | 804 | |||
BC059022.1 | - | 4 | 1292 | 6415235 |