Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256107.1 | NP_001243036.1 | 13 | 2483 | ||
NM_001256108.2 | NP_001243037.2 | 13 | 2303 | |||
NM_145124.3 | NP_660106.2 | 12 | 2387 | |||
GenBank mRNA | AB072336.1 | - | 12 | 1949 | ||
AK084791.1 | - | 5 | 685 | |||
AK147940.1 | - | 13 | 2248 | |||
AK159611.1 | - | 13 | 2200 | |||
AK183257.1 | - | 2 | 281 | |||
AK219099.1 | - | 2 | 193 | |||
AY974090.1 | - | 11 | 1684 | |||
BC058086.1 | - | 10 | 1971 | |||
BC141133.1 | - | 10 | 1776 | 9055745 | ||
BC145441.1 | - | 11 | 1872 |