Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Got1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029674.1 | NP_083950.1 | 5 | 605 | ||
GenBank mRNA | AK006984.1 | - | 5 | 592 | ||
BC052754.1 | - | 5 | 605 | 6533656 |