Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029701.1 | NP_083977.1 | 4 | 460 | ||
GenBank mRNA | AK007432.1 | - | 4 | 439 | ||
AK028314.1 | - | 4 | 422 | |||
AK048051.1 | - | 4 | 446 | |||
BC019390.1 | - | 4 | 399 | |||
BC054817.1 | - | 4 | 425 | 5375997 |