Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsper3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252487.1 | NP_001239416.1 | 3 | 542 | ||
NM_001252488.1 | NP_001239417.1 | 3 | 389 | |||
NM_029772.4 | NP_084048.1 | 3 | 503 | |||
GenBank mRNA | AK014942.1 | - | 3 | 504 | ||
AY263400.1 | - | 3 | 517 | |||
BC089518.1 | - | 642 | 6774298 | |||
HM179540.1 | - | 3 | 459 | |||
HM179541.1 | - | 3 | 306 |