Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Memo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133771.2 | NP_598532.1 | 2 | 219 | ||
GenBank mRNA | AF363447.1 | - | 3 | 281 | ||
AK053089.1 | - | 2 | 197 | |||
AK159560.1 | - | 2 | 183 | |||
BC013819.1 | - | 2 | 236 | 3489363 | ||
BF466279.1 | - | 2 | 219 |