Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256115.1 | NP_001243044.1 | 10 | 1152 | ||
NM_001256116.1 | NP_001243045.1 | 9 | 1008 | |||
NM_001256117.1 | NP_001243046.1 | 8 | 894 | |||
NM_001256118.1 | NP_001243047.1 | 6 | 732 | |||
NM_024451.2 | NP_077771.1 | 11 | 1263 | |||
GenBank mRNA | AK014585.1 | - | 8 | 892 | ||
AK036187.1 | - | 10 | 1129 | |||
AK160281.1 | - | 11 | 1221 | |||
AK167686.1 | - | 11 | 1235 | |||
AY682989.1 | - | 9 | 988 | |||
BC047928.1 | - | 11 | 1230 | 5321879 | ||
BC048156.1 | - | 11 | 1232 | 5698402 | ||
HQ402597.1 | - | 6 | 712 |