Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175256.5 | NP_780465.4 | 14 | 3202 | ||
GenBank mRNA | AK043598.1 | - | 3 | 1440 | ||
BC072651.1 | - | 6 | 512 | |||
BC076596.1 | - | 12 | 2481 | 30645937 |