Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175259.4 | NP_780468.1 | 2 | 205 | ||
GenBank mRNA | AK020369.1 | - | 2 | 223 | ||
AK039457.1 | - | 2 | 216 | |||
BC044813.1 | - | 2 | 205 | 4950177 |