Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prelid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029942.1 | NP_084218.1 | 1, 2 | 93 | ||
GenBank mRNA | AK021193.1 | - | 1, 2 | 119 | ||
BC119251.1 | - | 1, 2 | 93 | 8734000 | ||
BC120711.1 | - | 1, 2 | 93 | 40129634 | ||
BC145035.1 | - | 1, 2 | 93 | 9053583 |