Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrnp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290665.1 | NP_001277594.1 | 5 | 1298 | ||
NM_001290666.1 | NP_001277595.1 | 5 | 920 | |||
NM_153409.5 | NP_700458.3 | 7 | 1342 | |||
NM_178634.2 | NP_848749.2 | 6 | 1180 | |||
GenBank mRNA | AB091688.1 | - | 4 | 984 | ||
AK039150.1 | - | 7 | 1268 | |||
AK082649.1 | - | 6 | 1105 | |||
AK158873.1 | - | 5 | 1300 | |||
BC055701.1 | - | 5 | 832 | 6413559 | ||
EF210820.1 | - | 7 | 1314 |