Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029983.5 | NP_084259.1 | 3, 4 | 427 | ||
GenBank mRNA | AF287467.1 | - | 3, 4 | 478 | ||
AF434990.1 | - | 3, 4 | 470 | |||
AK020837.1 | - | 1, 2 | 84 | |||
AK030877.1 | - | 3, 4 | 428 | |||
AK088672.1 | - | 3, 4 | 554 | |||
AK138709.1 | - | 3, 4 | 428 | |||
BC052655.1 | - | 3, 4 | 526 | 30040401 |