Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133922.3 | NP_598683.3 | 12 | 1927 | ||
GenBank mRNA | AK029235.1 | - | 3 | 373 | ||
AK040525.1 | - | 12 | 1929 | |||
AK128931.1 | - | 12 | 1878 | |||
AK162934.1 | - | 12 | 1635 | |||
AK173281.1 | - | 12 | 3631 | |||
AK197252.1 | - | 3 | 275 | |||
BC066816.1 | - | 12 | 1847 | 30084911 |