Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msantd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145609.1 | NP_663584.1 | 2 | 1096 | ||
GenBank mRNA | AK032884.1 | - | 3 | 1147 | ||
AK132023.1 | - | 3 | 1132 | |||
BC014729.1 | - | 2 | 1096 | 4461077 |