Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153115.1 | NP_694755.1 | 2 | 149 | ||
GenBank mRNA | AB089183.1 | - | 2 | 149 | ||
AF384173.1 | - | 2 | 112 | |||
AK020333.1 | - | 2 | 153 | |||
AY552529.1 | - | 117 | ||||
BC089530.1 | - | 2 | 149 | |||
BC120808.1 | - | 2 | 114 | 40129731 | ||
BC120810.1 | - | 2 | 114 | 40129733 |