Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFYC SIRNA Assays ›
Gene Symbol:
NFYC
LOC494127
Gene Name:
nuclear transcription factor Y subunit gamma
NFYC pseudogene
Gene Aliases:
CBF-C, CBFC, H1TF2A, HAP5, HSM, NF-YC
-
Chromosome Location:
Chr.1: 40691570 - 40771603 on Build GRCh38
Chromosome Location:
Chr.9: 87083886 - 87085225 on Build GRCh38
UniGene ID:
Hs.713051
Hs.626316
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s9536
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFYC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142587.1
NP_001136059.1
6
841
NM_001142588.1
NP_001136060.1
6
841
NM_001142589.1
NP_001136061.1
5
727
NM_001142590.1
NP_001136062.1
11
1218
NM_001308114.1
NP_001295043.1
5
543
NM_001308115.1
NP_001295044.1
6
837
NM_014223.4
NP_055038.2
6
841
XM_005270894.1
XP_005270951.1
6
837
XM_005270895.3
XP_005270952.3
6
693
XM_006710658.2
XP_006710721.2
6
693
XM_006710660.2
XP_006710723.2
6
693
XM_006710661.1
XP_006710724.1
6
722
XM_006710662.2
XP_006710725.2
6
693
XM_011541516.1
XP_011539818.1
6
642
XM_011541517.1
XP_011539819.1
6
693
XM_017001364.1
XP_016856853.1
6
862
XM_017001365.1
XP_016856854.1
6
722
XM_017001366.1
XP_016856855.1
6
796
XM_017001367.1
XP_016856856.1
6
837
XM_017001368.1
XP_016856857.1
6
837
XM_017001369.1
XP_016856858.1
7
1194
GenBank mRNA
AB208975.1
-
6
668
AF191744.1
-
5
542
AK000346.1
-
6
794
AK127677.1
-
11
1213
AK300774.1
-
6
644
AK301215.1
-
3
443
AK301385.1
-
5
611
BC005003.1
-
6
690
3502903
BT020081.1
-
5
535
D85425.1
-
6
618
U62296.1
-
5
535
U78774.1
-
7
709
Z74792.1
-
7
720
Gene Symbol: LOC494127
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_036691.1
1
599
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein folding
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
transcription factor binding
transcription regulatory region DNA binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0