Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030093.3 | NP_084369.2 | 4 | 470 | ||
GenBank mRNA | AA199179.1 | - | 1 | 39 | ||
AK019452.1 | - | 4 | 471 | |||
BC028766.1 | - | 4 | 411 | 1244556 |