Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020610.1 | NP_065635.1 | 2 | 350 | ||
GenBank mRNA | AK020731.1 | - | 2 | 352 | ||
AK083130.1 | - | 2 | 352 | |||
BC072641.1 | - | 3 | 363 | 6839519 |