Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Troap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162506.1 | NP_001155978.1 | 8 | 973 | ||
NM_030159.1 | NP_084435.1 | 8 | 998 | |||
GenBank mRNA | AK050381.1 | - | 8 | 994 | ||
AK077509.1 | - | 8 | 963 | |||
BC080746.1 | - | 8 | 958 | |||
BC100405.1 | - | 8 | 927 | |||
BC145227.1 | - | 8 | 1371 | 9053779 |