Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175279.3 | NP_780488.2 | 1 | 1053 | ||
GenBank mRNA | AK019489.1 | - | 1 | 500 | ||
AK031660.1 | - | 1 | 1054 | |||
AK046433.1 | - | 1 | 1049 | |||
BC028854.1 | - | 1 | 629 | |||
BC125291.1 | - | 1 | 869 | 40129806 | ||
BC125293.1 | - | 1 | 869 | 40129808 |