Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005385.3 | NP_005376.2 | 13 | 3382 | ||
XM_005265173.1 | XP_005265230.1 | 13 | 3377 | |||
XM_006713173.2 | XP_006713236.1 | 14 | 3399 | |||
XM_011533747.2 | XP_011532049.1 | 14 | 6711 | |||
XM_011533750.2 | XP_011532052.1 | 15 | 3393 | |||
XM_017006475.1 | XP_016861964.1 | 14 | 3381 | |||
XM_017006476.1 | XP_016861965.1 | 8 | 7924 | |||
XM_017006477.1 | XP_016861966.1 | 8 | 4578 | |||
XM_017006478.1 | XP_016861967.1 | 8 | 4555 | |||
XM_017006479.1 | XP_016861968.1 | 7 | 5311 | |||
XM_017006480.1 | XP_016861969.1 | 15 | 3389 | |||
GenBank mRNA | BX957215.1 | - | 5 | 4558 | ||
BX957218.1 | - | 15 | 3407 | |||
CR749415.1 | - | 5 | 4558 | |||
CR749571.1 | - | 15 | 3407 | |||
L04288.2 | - | 13 | 3423 |