Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030176.2 | NP_084452.2 | 3 | 1042 | ||
GenBank mRNA | AK019166.1 | - | 2 | 1368 | ||
AK081234.1 | - | 3 | 1042 | |||
AK170267.1 | - | 2 | 1342 | |||
AK220503.1 | - | 1342 | ||||
BC139310.1 | - | 2 | 977 | 8862332 | ||
BC139311.1 | - | 2 | 977 | 8862333 |