Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030225.4 | NP_084501.1 | 13 | 1099 | ||
GenBank mRNA | AK005477.1 | - | 1 | 76 | ||
AK019713.2 | - | 1096 | ||||
AK054053.1 | - | 13 | 1053 | |||
AK149664.1 | - | 13 | 1075 | |||
AK158877.1 | - | 13 | 1100 | |||
AK168570.1 | - | 13 | 1075 | |||
AK169943.1 | - | 13 | 1053 | |||
AK197945.1 | - | 4 | 302 | |||
AK211646.1 | - | 3 | 271 | |||
AK214016.1 | - | 4 | 356 | |||
AK220022.1 | - | 4 | 302 | |||
BC006702.1 | - | 13 | 1036 | 3583936 | ||
BC024066.1 | - | 4 | 486 | |||
CT010197.1 | - | 13 | 1035 |