Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133779.2 | NP_598540.2 | 8 | 1052 | ||
GenBank mRNA | AK019717.2 | - | 8 | 1042 | ||
AK044499.1 | - | 8 | 1036 | |||
AK049171.1 | - | 8 | 1046 | |||
AK049990.1 | - | 8 | 1045 | |||
AK090020.1 | - | 8 | 1044 | |||
AK153462.1 | - | 8 | 1038 | |||
AK153464.1 | - | 8 | 1038 | |||
AK155057.1 | - | 8 | 1039 | |||
AK157226.1 | - | 8 | 1032 | |||
AK162573.1 | - | 6 | 1250 | |||
BC016520.1 | - | 9 | 1106 | |||
BC034175.1 | - | 8 | 1028 | 5026752 |