Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163750.1 | NP_001157222.1 | 6 | 1154 | ||
NM_001163751.1 | NP_001157223.1 | 6 | 809 | |||
NM_027141.2 | NP_081417.2 | 6 | 830 | |||
GenBank mRNA | AF403038.1 | - | 6 | 830 | ||
AK009325.1 | - | 6 | 830 | |||
AK076170.1 | - | 6 | 1106 | |||
AK082768.1 | - | 6 | 753 | |||
AK156579.1 | - | 5 | 1167 | |||
AK167474.1 | - | 5 | 1147 | |||
AK176934.1 | - | 2 | 169 | |||
AK183595.1 | - | 2 | 169 | |||
AK191468.1 | - | 2 | 170 | |||
AK208055.1 | - | 2 | 169 | |||
AK210660.1 | - | 2 | 169 | |||
AK212011.1 | - | 1 | 244 | |||
AK214702.1 | - | 1 | 244 | |||
AK216422.1 | - | 1 | 244 | |||
AK220234.1 | - | 6 | 1092 | |||
BC026523.1 | - | 6 | 1495 | |||
BC072556.1 | - | 6 | 1115 | 5691114 |