Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ublcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024475.5 | NP_077795.2 | 7 | 733 | ||
GenBank mRNA | AK043952.1 | - | 7 | 640 | ||
AK050031.1 | - | 7 | 644 | |||
AK147744.1 | - | 7 | 681 | |||
AK153998.1 | - | 7 | 645 | |||
BC056652.1 | - | 3 | 234 | |||
BC083331.1 | - | 5 | 394 | |||
BC085111.1 | - | 6 | 636 | 30735741 | ||
BC094051.1 | - | 1 | 39 | |||
BC106093.1 | - | 6 | 610 | 5098953 |