Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030556.2 | NP_085033.2 | 6 | 1385 | ||
GenBank mRNA | AF271634.1 | - | 6 | 1385 | ||
AK079566.1 | - | 3 | 304 | |||
AK098112.1 | - | 6 | 1376 | |||
BC109154.1 | - | 6 | 1368 | 40047017 | ||
BC109155.1 | - | 5 | 1238 | 40047019 |