Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030563.2 | NP_085040.2 | 5 | 2391 | ||
GenBank mRNA | AK147522.1 | - | 5 | 2392 | ||
AK172981.1 | - | 5 | 2373 | |||
BC004022.1 | - | 2 | 938 | |||
BC089324.1 | - | 4 | 1556 | |||
BC111031.1 | - | 2 | 836 |