Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030599.4 | NP_085102.4 | 6 | 1677 | ||
GenBank mRNA | AK030702.1 | - | 6 | 1626 | ||
BC055379.1 | - | 6 | 1676 | 30076215 | ||
BC100580.1 | - | 6 | 1664 | 30074482 |