Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maged2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199246.1 | NP_001186175.1 | 12 | 1672 | ||
NM_030700.2 | NP_109625.1 | 12 | 1695 | |||
GenBank mRNA | AF319976.1 | - | 11 | 1562 | ||
AJ277114.1 | - | 12 | 1638 | |||
AK131598.1 | - | 7 | 734 | |||
AK138441.1 | - | 12 | 1612 | |||
AK169006.1 | - | 12 | 1644 | |||
AK189405.1 | - | 1 | 148 | |||
AK206158.1 | - | 1 | 148 | |||
BC023319.1 | - | 10 | 2140 | |||
BC058581.1 | - | 12 | 1651 | 6491028 |