Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000625.4 | NP_000616.3 | 6 | 766 | ||
XM_011524859.2 | XP_011523161.1 | 6 | 792 | |||
XM_011524860.2 | XP_011523162.1 | 5 | 585 | |||
XM_011524861.1 | XP_011523163.1 | 6 | 764 | |||
XM_011524862.1 | XP_011523164.1 | 4 | 363 | |||
GenBank mRNA | AB022318.1 | - | 5 | 502 | ||
AF068236.1 | - | 6 | 736 | |||
AK304252.1 | - | 6 | 764 | |||
BC130283.1 | - | 6 | 686 | 40146314 | ||
BC144126.1 | - | 6 | 686 | 9052646 | ||
D26525.1 | - | 5 | 516 | |||
L09210.1 | - | 6 | 708 | |||
L24553.1 | - | 5 | 578 | |||
U05810.1 | - | 6 | 696 | |||
U20141.1 | - | 5 | 502 | |||
U31511.1 | - | 6 | 607 | |||
X73029.1 | - | 6 | 727 |