Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1xr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030732.3 | NP_109657.2 | 5, 6 | 836 | ||
GenBank mRNA | AF268195.1 | - | 5, 6 | 617 | ||
AK029595.1 | - | 5, 6 | 635 | |||
AK033347.1 | - | 5, 6 | 706 | |||
AK036064.1 | - | 6, 7 | 947 | |||
AK136255.1 | - | 5, 6 | 658 |