Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031179.2 | NP_112456.2 | 25 | 3896 | ||
GenBank mRNA | AB037890.1 | - | 25 | 3921 | ||
AK048521.1 | - | 3 | 1059 |