Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4x1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003947.1 | NP_001003947.1 | 6, 7 | 781 | ||
GenBank mRNA | AJ786769.1 | - | 6, 7 | 781 | ||
AK038526.1 | - | 757 | ||||
BC113125.1 | - | 6, 7 | 780 |