Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnnm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031396.2 | NP_113573.2 | 7, 8 | 2356 | ||
GenBank mRNA | AF202994.1 | - | 7, 8 | 1582 | ||
DQ885890.5 | - | 7, 8 | 1252 | |||
EF041508.1 | - | 8, 9 | 1315 |