Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109909.1 | NP_001103379.1 | 15 | 2227 | ||
NM_001109910.1 | NP_001103380.1 | 15 | 2227 | |||
NM_031405.2 | NP_113582.1 | 15 | 2227 | |||
GenBank mRNA | AK144300.1 | - | 15 | 2219 | ||
AK150310.1 | - | 4 | 533 | |||
AK152710.1 | - | 4 | 533 | |||
BC019117.1 | - | 6 | 769 | |||
BC066831.1 | - | 15 | 2186 | 30471339 | ||
CB195376.1 | - | 1 | 138 |