Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114386.1 | NP_001107858.1 | 14 | 1505 | ||
NM_031881.2 | NP_114087.2 | 13 | 1554 | |||
GenBank mRNA | AF277232.1 | - | 13 | 1187 | ||
AK004969.1 | - | 13 | 1247 | |||
AK031842.1 | - | 12 | 1175 | |||
AK042621.1 | - | 13 | 1128 | |||
AK163497.1 | - | 11 | 975 | |||
AK183000.1 | - | 3 | 195 | |||
BC039746.1 | - | 13 | 1198 | 3157137 | ||
BC071210.1 | - | 13 | 1175 | 4972426 |