Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002515.2 | NP_002506.2 | 2 | 538 | ||
NM_006489.2 | NP_006480.2 | 2 | 538 | |||
NM_006491.2 | NP_006482.1 | 2 | 538 | |||
XM_017021342.1 | XP_016876831.1 | 2 | 594 | |||
XM_017021343.1 | XP_016876832.1 | 2 | 594 | |||
XM_017021344.1 | XP_016876833.1 | 2 | 303 | |||
XM_017021345.1 | XP_016876834.1 | 2 | 752 | |||
XM_017021346.1 | XP_016876835.1 | 2 | 752 | |||
XM_017021347.1 | XP_016876836.1 | 2 | 633 | |||
XM_017021349.1 | XP_016876838.1 | 2 | 753 | |||
GenBank mRNA | AK226114.1 | - | 2 | 255 | ||
AK289639.1 | - | 2 | 224 | |||
AK291022.1 | - | 2 | 361 | |||
AK291468.1 | - | 2 | 239 | |||
AK301526.1 | - | 2 | 220 | |||
BC075038.2 | - | 2 | 243 | 30915356 | ||
BC075039.2 | - | 2 | 244 | 30915572 | ||
CN409914.1 | - | 2 | 541 | |||
U04840.1 | - | 2 | 280 |