Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032541.1 | NP_115930.1 | 1, 2 | 128 | ||
GenBank mRNA | AF297664.1 | - | 1, 2 | 128 | ||
AF503444.1 | - | 1, 2 | 128 | |||
BC021587.1 | - | 1, 2 | 111 | 5097807 |
Gene Symbol: Hamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183257.3 | NP_899080.1 | 1, 2 | 143 | ||
GenBank mRNA | AK007975.1 | - | 1, 2 | 133 | ||
AK008993.1 | - | 1, 2 | 144 | |||
AY232841.1 | - | 1, 2 | 132 | |||
BC086895.1 | - | 1, 2 | 140 | 6765293 |