Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Emc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033146.1 | NP_149158.1 | 3 | 447 | ||
GenBank mRNA | AB054001.1 | - | 3 | 493 | ||
AK005154.1 | - | 3 | 447 | |||
AK144394.1 | - | 3 | 475 | |||
AK154475.1 | - | 3 | 496 | |||
BC024704.1 | - | 3 | 423 | 4195108 |