Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002518.3 | NP_002509.2 | 9, 10 | 1097 | ||
XM_005263953.2 | XP_005264010.1 | 9, 10 | 1008 | |||
XM_005263959.2 | XP_005264016.1 | 9, 10 | 1008 | |||
XM_005263961.4 | XP_005264018.1 | 7, 8 | 902 | |||
XM_011511242.2 | XP_011509544.1 | 8, 9 | 975 | |||
XM_011511243.2 | XP_011509545.1 | 9, 10 | 1008 | |||
XM_017004214.1 | XP_016859703.1 | 9, 10 | 1008 | |||
XM_017004215.1 | XP_016859704.1 | 9, 10 | 1008 | |||
XM_017004216.1 | XP_016859705.1 | 9, 10 | 1008 | |||
GenBank mRNA | AK226093.1 | - | 4, 5 | 425 | ||
AK294733.1 | - | 9, 10 | 1008 | |||
BC028107.1 | - | 8, 9 | 840 | |||
BC051351.1 | - | 9, 10 | 1061 | 6538324 | ||
BC072383.1 | - | 9, 10 | 1064 | 6371187 | ||
BM724652.1 | - | 2, 3 | 192 | |||
KJ534892.1 | - | 8, 9 | 811 | |||
U51625.1 | - | 8, 9 | 846 | |||
U77970.1 | - | 9, 10 | 1097 |