Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110827.2 | NP_001104297.1 | 3 | 780 | ||
NM_001110828.2 | NP_001104298.1 | 2 | 322 | |||
NM_001110829.2 | NP_001104299.1 | 2 | 322 | |||
NM_001110830.2 | NP_001104300.1 | 2 | 322 | |||
NM_001286417.1 | NP_001273346.1 | 2 | 322 | |||
NM_001286418.1 | NP_001273347.1 | 2 | 322 | |||
NM_001286419.1 | NP_001273348.1 | 2 | 322 | |||
NM_053104.6 | NP_444334.3 | 3 | 780 | |||
NM_175387.3 | NP_780596.1 | 3 | 780 | |||
GenBank mRNA | AF229055.1 | - | 2 | 265 | ||
AF387322.1 | - | 2 | 66 | |||
AK077594.1 | - | 3 | 656 | |||
AK139555.1 | - | 4 | 750 | |||
AY659951.1 | - | 2 | 66 | |||
AY659952.1 | - | 2 | 66 | |||
AY659953.1 | - | 2 | 66 | |||
AY904025.1 | - | 4 | 216 | |||
BC002124.1 | - | 2 | 160 | 3494630 | ||
BC027263.1 | - | 3 | 332 | 3661748 | ||
BY238482.1 | - | 2 | 333 | |||
BY247199.1 | - | 2 | 324 | |||
DQ017388.1 | - | 2 | 66 | |||
DQ017389.1 | - | 2 | 66 |