Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161667.1 | NP_001155139.1 | 8 | 1006 | ||
NM_053115.2 | NP_444345.2 | 8 | 1131 | |||
GenBank mRNA | AJ238492.1 | - | 8 | 1048 | ||
AK144243.1 | - | 8 | 1000 | |||
BC021339.1 | - | 8 | 1010 | 5052291 |