Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabarapl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026693.5 | NP_080969.2 | 3, 4 | 387 | ||
GenBank mRNA | AF190644.1 | - | 3, 4 | 386 | ||
AK002596.1 | - | 3, 4 | 367 | |||
AK049819.1 | - | 369 | ||||
AK134873.1 | - | 388 | ||||
BC026798.1 | - | 3, 4 | 347 | 4036425 | ||
BC081436.1 | - | 3, 4 | 372 | 6814986 |