Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053177.1 | NP_444407.1 | 6, 7 | 862 | ||
GenBank mRNA | AF302009.1 | - | 6, 7 | 859 | ||
AK028385.1 | - | 6, 7 | 839 | |||
AK034471.1 | - | 6, 7 | 873 | |||
AK038594.1 | - | 6, 7 | 875 | |||
AK049606.1 | - | 6, 7 | 863 | |||
BC005651.1 | - | 6, 7 | 862 | 3257440 |